Assay Details
| Gene Symbol: Gm16630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK158926.1 | - | 2 - 3 | 853 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 746 | Zfp746 |
2810407L07Rik, AI317225, Znf746 |
-66.7kb | Sense | Chr. 6 - 48062381 - 48086990 on NCBI Build GRCm38 |
| Downstream | KRAB-A domain containing 1 | Krba1 |
A930040G15Rik, AI448780 |
236.4kb | Anti-sense | Chr. 6 - 48395586 - 48419855 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm16630
Genomic Map