Assay Details
| Gene Symbol: Slc31a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK172012.1 | - | 1 | 1950 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FK506 binding protein 15 | Fkbp15 |
BB131447, C430014M02Rik, FKBO133, FKBP-133, FKBP-15, FKBP133, mKIAA0674 |
-110.0 | Anti-sense | Chr. 4 - 62300342 - 62360591 on NCBI Build GRCm38 |
| Downstream | cell division cycle 26 | Cdc26 | 2010012C09Rik | 30.3kb | Anti-sense | Chr. 4 - 62393812 - 62408640 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc31a1
Genomic Map