Assay Details
| Gene Symbol: Gm26515 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK131823.1 | - | 1 - 2 | 277 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | pseudouridine synthase 1 | Pus1 |
A730013B20Rik, MPUS1, mPus1p |
Anti-sense | Chr. 5 - 110773667 - 110781056 on NCBI Build GRCm38 | |
| Downstream | unc-51 like kinase 1 | Ulk1 |
AU041434, Unc51.1, mKIAA0722 |
3.4kb | Anti-sense | Chr. 5 - 110784489 - 110810118 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm26515
Genomic Map