Assay Details
| Gene Symbol: Gm16755 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK131899.1 | - | 2 - 3 | 228 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitochondrial ribosomal protein L12 | Mrpl12 |
0610034O11Rik, 1500031N16Rik, MRP-L12, Rpml12 |
Anti-sense | Chr. 11 - 120484669 - 120488754 on NCBI Build GRCm38 | |
| Downstream |
solute carrier family 25 (mitochondrial carrier, dicarboxylate transporter), member 10 |
Slc25a10 | Dic | 3.7kb | Anti-sense | Chr. 11 - 120491837 - 120501161 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm16755
Genomic Map