Assay Details
| Gene Symbol: Gm11351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045062.1 | 3 - 4 | 321 | 72 | ||
| GenBank mRNA | AK132076.1 | - | 3 - 4 | 321 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neurensin 1 | Nrsn1 |
Neurensin-1, Neuro-p24, Vmp |
-651.0kb | Sense | Chr. 13 - 25252039 - 25270070 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700092E19 gene | 1700092E19Rik | 360.6kb | Sense | Chr. 13 - 26283159 - 26312406 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 32494 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 734755 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149829 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350559 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 87395 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 42754 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 203578 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 534303 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515632 | AB183189.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515632 | AF012190.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515632 | AF296659.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515632 | AK041407.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515632 | AK042313.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515632 | AK134087.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515632 | X02933.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515632 | X02939.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515632 | X02967.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515632 | XM_001479551.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515632 | XM_001479646.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515632 | XM_903593.2 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515632 | XM_908597.2 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773982 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618340 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 601305 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 702699 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 148350 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 168707 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 51091 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 61328 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 28449 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07815636 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07815733 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07815715 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07815688 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07815715 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07815636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895156 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28894968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10848125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28894965 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28894966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28894951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10656101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895216 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28894946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10830244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895149 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28894949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895227 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10671456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10522512 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10830249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10622538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10671437 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895117 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10709389 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895153 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28894969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10942137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895208 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11078313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28894961 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28894956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10722798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28894942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10771292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10556182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28894950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28894947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28894964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28894962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895215 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28894953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10671432 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28894944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895202 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28894952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28895042 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR1300010449.mm.1 | NR_045062.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010449.mm.1 | NR_045062.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010449.mm.1 | TC1300001679.mm.1 | MTA TC |

Gm11351
Genomic Map