Assay Details
| Gene Symbol: Gm29230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK132084.1 | - | 2 - 3 | 262 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 37548 |
Gm37548 | -170.0kb | Sense | Chr. 3 - 50883237 - 50888892 on NCBI Build GRCm38 | |
| Downstream | CCR4 carbon catabolite repression 4-like (S. cerevisiae) | Ccrn4l |
AU043840, Ccr4, nocturnin |
161.9kb | Anti-sense | Chr. 3 - 51224447 - 51251654 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm29230
Genomic Map