Assay Details
| Gene Symbol: Gm34639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK136628.1 | - | 3 - 4 | 322 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 2610307P16 gene | 2610307P16Rik | AI666457 | -8.4kb | Sense | Chr. 13 - 28460034 - 28885422 on NCBI Build GRCm38 |
| Downstream | SRY (sex determining region Y)-box 4 | Sox4 |
AA682046, Sox-4 |
56.4kb | Sense | Chr. 13 - 28950716 - 28953682 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm34639
Genomic Map