Assay Details
| Gene Symbol: Gm26550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK136710.1 | - | 4 - 5 | 473 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
pleckstrin homology-like domain, family B, member 3 |
Phldb3 |
EG232970, ENSMUSG00000061511, Gm10102 |
-1.8kb | Anti-sense | Chr. 7 - 24610272 - 24629297 on NCBI Build GRCm38 |
| Downstream | Ly6/Plaur domain containing 3 | Lypd3 |
2310061G07Rik, C4.4a |
2.6kb | Anti-sense | Chr. 7 - 24636570 - 24641118 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm26550
Genomic Map