Assay Details
| Gene Symbol: Gm21284 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_078345.1 | 3 - 4 | 452 | 75 | ||
| GenBank mRNA | AK137133.1 | - | 3 - 4 | 453 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | STAM binding protein | Stambp |
5330424L14Rik, 5730422L11Rik, AW107289, Amsh, mKIAA4198 |
-294.0 | Anti-sense | Chr. 6 - 83543206 - 83575770 on NCBI Build GRCm38 |
| Downstream |
C-type lectin domain family 4, member f |
Clec4f |
Clecsf13, D18063, KUCR_MOUSE, Kclr |
64.9kb | Anti-sense | Chr. 6 - 83644542 - 83656116 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm21284
Genomic Map