Assay Details
| Gene Symbol: Gm20465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK138059.1 | - | 1 - 2 | 220 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | C-terminal binding protein 1 | Ctbp1 |
BARS, CtBP1-L, CtBP1-S, CtBP3/BARS, D4S115h, D5H4S115, D5H4S115E |
-23.1kb | Sense | Chr. 5 - 33247723 - 33275004 on NCBI Build GRCm38 |
| Downstream | macrophage erythroblast attacher | Maea |
1110030D19Rik, EMP, Gid9 |
24.4kb | Anti-sense | Chr. 5 - 33335572 - 33373294 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm20465
Genomic Map