Assay Details
| Gene Symbol: Gm15958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK138348.1 | - | 371 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 694 | Mir694 |
Mirn694, mir-694, mmu-mir-694 |
-71.9kb | Anti-sense | Chr. 18 - 66219264 - 66219333 on NCBI Build GRCm38 |
| Downstream | phorbol-12-myristate-13-acetate-induced protein 1 | Pmaip1 | Noxa | 146.3kb | Sense | Chr. 18 - 66458604 - 66465558 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm15958
Genomic Map