Assay Details
| Gene Symbol: Gm41505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK142264.1 | - | 189 | 112 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
potassium inwardly-rectifying channel, subfamily J, member 6 |
Kcnj6 |
BIR1, GIRK2, KATP2, KCNJ7, Kir3.2, weaver, wv |
Anti-sense | Chr. 16 - 94748683 - 94997696 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 31641 |
Gm31641 | 435.2kb | Anti-sense | Chr. 16 - 95184537 - 95263455 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm41505
Genomic Map