Assay Details
| Gene Symbol: Gm17308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK142469.1 | - | 386 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1110015O18 gene | 1110015O18Rik | -275.4kb | Sense | Chr. 3 - 4798708 - 4814911 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 2700069I18 gene | 2700069I18Rik | 84.5kb | Anti-sense | Chr. 3 - 5177824 - 5220823 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm17308
Genomic Map