Assay Details
| Gene Symbol: Gm35521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK145215.1 | - | 2 - 3 | 230 | 110 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tolloid-like | Tll1 |
Tll, Tll-1 |
-202.7kb | Sense | Chr. 8 - 64014150 - 64205993 on NCBI Build GRCm38 |
| Downstream | microRNA 710 | Mir710 |
Mirn710, mmu-mir-710 |
102.3kb | Sense | Chr. 8 - 64514332 - 64514441 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm35521
Genomic Map