Assay Details
| Gene Symbol: Gm19461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037984.1 | 4 | 3011 | 96 | ||
| GenBank mRNA | AK145464.1 | - | 5 | 2719 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
pleckstrin homology domain containing, family A member 6 |
Plekha6 |
9530028L01Rik, AU042621, Pepp3 |
Anti-sense | Chr. 1 - 133163771 - 133303435 on NCBI Build GRCm38 | |
| Downstream | golgi transport 1 homolog A (S. cerevisiae) | Golt1a |
0610012C01Rik, Kiss1, metastatin |
59.7kb | Anti-sense | Chr. 1 - 133309823 - 133323026 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm19461
Genomic Map