Assay Details
| Gene Symbol: Psen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK148390.1 | - | 1 | 1447 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RNA binding motif protein 25 | Rbm25 |
2600011C06Rik, 2610015J01Rik, A130095G20Rik, AI159652, AL023075, AU043498, RNPC7 |
-53.7kb | Sense | Chr. 12 - 83632204 - 83683127 on NCBI Build GRCm38 |
| Downstream |
papilin, proteoglycan-like sulfated glycoprotein |
Papln | E030033C16Rik | 24.6kb | Sense | Chr. 12 - 83762992 - 83794875 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Psen1
Genomic Map