Assay Details
| Gene Symbol: Dnaja1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164671.1 | NP_001158143.1 | 9 | 4000 | 80 | |
| NM_001164672.1 | NP_001158144.1 | 9 | 3857 | 80 | ||
| NM_008298.5 | NP_032324.1 | 9 | 3951 | 80 | ||
| GenBank mRNA | AK163758.1 | - | 1 | 1652 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 207 | Mir207 |
Mirn207, mmu-mir-207 |
-10.9kb | Sense | Chr. 4 - 40722917 - 40722995 on NCBI Build GRCm38 |
| Downstream |
UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 1 |
B4galt1 |
B-1,4-GalT, B-1,4-GalT1, GalT, Ggtb, Ggtb2, b4Gal-T1, beta-1,4-GalT, beta-1,4-GalT1, beta4Gal-T1 |
67.4kb | Anti-sense | Chr. 4 - 40804582 - 40854537 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dnaja1
Genomic Map