Assay Details
| Gene Symbol: Gm4117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK087351.1 | - | 130 | 103 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | X-ray repair complementing defective repair in Chinese hamster cells 4 | Xrcc4 |
2310057B22Rik, AW413319, AW545101 |
Anti-sense | Chr. 13 - 89848904 - 90089608 on NCBI Build GRCm38 | |
| Downstream | transmembrane protein 167 | Tmem167 |
0610041E09Rik, 5730424F14Rik, AU041184, AW537806, ENSMUSG00000060669, Gm10085, Tmem167a |
202.6kb | Sense | Chr. 13 - 90089667 - 90114921 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm4117
Genomic Map