Assay Details
| Gene Symbol: Gm15350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045775.1 | 3 | 446 | 85 | ||
| GenBank mRNA | AK006628.1 | - | 3 | 448 | 85 | |
| AK018911.1 | - | 3 | 447 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 15348 | Gm15348 |
ENSMUSG00000074469, OTTMUSG00000020606 |
-109.2kb | Sense | Chr. 8 - 12706944 - 12719127 on NCBI Build GRCm38 |
| Downstream | mcf.2 transforming sequence-like | Mcf2l |
C130040G20Rik, Dbs, Ost, mKIAA0362 |
45.2kb | Anti-sense | Chr. 8 - 12873811 - 13020509 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm15350
Genomic Map