Assay Details
| Gene Symbol: Gm15834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042008.1 | - | 1 | 748 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
origin recognition complex, subunit 2 |
Orc2 |
AU041563, Orc2l |
-35.0 | Anti-sense | Chr. 1 - 58462771 - 58505109 on NCBI Build GRCm38 |
| Downstream |
family with sequence similarity 126, member B |
Fam126b |
C130065N10Rik, D1Ertd53e, D630010C10 |
15.9kb | Anti-sense | Chr. 1 - 58522806 - 58586333 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm15834
Genomic Map