Assay Details
| Gene Symbol: Celf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043349.1 | - | 2 | 1849 | 111 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 10855 | Gm10855 | ENSMUSG00000075538 | -137.7kb | Sense | Chr. 2 - 6932547 - 6939595 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700061F12 gene | 1700061F12Rik | 2.1mb | Anti-sense | Chr. 2 - 9189515 - 9197484 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Celf2
Genomic Map