Assay Details
| Gene Symbol: Gm12996 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK078255.1 | - | 2 | 256 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | 4-hydroxyphenylpyruvate dioxygenase-like | Hpdl |
A830048M07Rik, BC034099, Gloxd1 |
-8.5kb | Anti-sense | Chr. 4 - 116819907 - 116821508 on NCBI Build GRCm38 |
| Downstream | zinc finger SWIM-type containing 5 | Zswim5 |
4933426E21Rik, AI503093, mKIAA1511 |
46.8kb | Sense | Chr. 4 - 116877402 - 116989105 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm12996
Genomic Map