Assay Details
| Gene Symbol: Arf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK082322.1 | - | 1 | 3549 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 2310033P09 gene | 2310033P09Rik | Mmtag2 | -4.9kb | Anti-sense | Chr. 11 - 59208361 - 59210736 on NCBI Build GRCm38 |
| Downstream |
wingless-type MMTV integration site family, member 3A |
Wnt3a |
Wnt-3a, vt |
28.6kb | Sense | Chr. 11 - 59248036 - 59290751 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Arf1
Genomic Map