Assay Details
| Gene Symbol: Gm17518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK090163.1 | - | 2 | 844 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MIS18 kinetochore protein homolog A (S. pombe) | Mis18a |
2610039C10Rik, 2810018N07Rik |
-949.0 | Anti-sense | Chr. 16 - 90719312 - 90727371 on NCBI Build GRCm38 |
| Downstream | melanocortin 2 receptor accessory protein | Mrap |
1110025G12Rik, Falp, ORF61 |
9.2kb | Sense | Chr. 16 - 90738324 - 90749776 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm17518
Genomic Map