Assay Details
| Gene Symbol: Mvb12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK132338.1 | - | 2 | 2252 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 9430024E24 gene | 9430024E24Rik | -46.6kb | Sense | Chr. 2 - 33714606 - 33718899 on NCBI Build GRCm38 | |
| Downstream |
non-protein coding RNA, repressor of NFAT |
Nron |
A630071L07Rik, Gm24353 |
39.6kb | Anti-sense | Chr. 2 - 33805816 - 33813591 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mvb12b
Genomic Map