Assay Details
| Gene Symbol: Gm36037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK138343.1 | - | 2 | 1352 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | heat shock protein 9 | Hspa9 |
74kDa, Csa, Grp75, Hsc74, Hsp74, Hsp74a, Hspa9a, Mortalin, Mot-2, Mot2, Mthsp70, Pbp74 |
-45.7kb | Anti-sense | Chr. 18 - 34937414 - 34954351 on NCBI Build GRCm38 |
| Downstream |
catenin (cadherin associated protein), alpha 1 |
Ctnna1 |
2010010M04Rik, AA517462, AI988031, Catna1 |
116.4kb | Sense | Chr. 18 - 35118912 - 35254775 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm36037
Genomic Map