Assay Details
| Gene Symbol: A230095H09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK138404.1 | - | 1 | 2252 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SH3/ankyrin domain gene 3 | Shank3 |
AI841104, SHANK3c-3, SHANK3c-4, Shank3b |
-2.7kb | Sense | Chr. 15 - 89499707 - 89560261 on NCBI Build GRCm38 |
| Downstream | acrosin prepropeptide | Acr | 1.2kb | Sense | Chr. 15 - 89567562 - 89578904 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A230095H09Rik
Genomic Map