Assay Details
| Gene Symbol: Gm13483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040361.1 | 3 | 357 | 91 | ||
| GenBank mRNA | AK142467.1 | - | 3 | 357 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
methylmalonic aciduria (cobalamin deficiency) cblD type, with homocystinuria |
Mmadhc |
2010311D03Rik, AI314967 |
-67.3kb | Anti-sense | Chr. 2 - 50279881 - 50296799 on NCBI Build GRCm38 |
| Downstream |
heat shock protein 90 alpha (cytosolic), class B member 1 pseudogene |
Gm13498 | OTTMUSG00000012426 | 544.7kb | Sense | Chr. 2 - 50909684 - 50911846 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm13483
Genomic Map