Assay Details
| Gene Symbol: Gm26644 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK144194.1 | - | 2 | 652 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
hypoxia-inducible factor 1, alpha subunit inhibitor |
Hif1an |
2310046M24Rik, A830014H24Rik, FIH, FIH1 |
-116.1kb | Anti-sense | Chr. 19 - 44562854 - 44576274 on NCBI Build GRCm38 |
| Downstream | paired box 2 | Pax2 |
Opdc, Pax-2 |
52.4kb | Anti-sense | Chr. 19 - 44745695 - 44837871 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm26644
Genomic Map