Assay Details
| Gene Symbol: AA692955 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK159351.1 | - | 1 | 447 | 199 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | bromodomain containing 4 | Brd4 |
Brd5, HUNK1, MCAP, WI-11513 |
-65.0 | Anti-sense | Chr. 17 - 32196272 - 32284722 on NCBI Build GRCm38 |
| Downstream | A kinase (PRKA) anchor protein 8 | Akap8 |
1200016A02Rik, AA673585, AKAP-8, AKAP95, AU015639 |
17.8kb | Anti-sense | Chr. 17 - 32303672 - 32321327 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

AA692955
Genomic Map