Assay Details
| Gene Symbol: Gm15169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK163197.1 | - | 2049 | 92 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small muscle protein, X-linked |
Smpx |
1010001C09Rik, Csl |
-4.8kb | Anti-sense | Chr. X - 157698973 - 157752591 on NCBI Build GRCm38 |
| Downstream | kelch-like 34 | Klhl34 | Gm388 | 60.3kb | Anti-sense | Chr. X - 157818435 - 157821060 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm15169
Genomic Map