Assay Details
| Gene Symbol: Gm12610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK166205.1 | - | 1245 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4) |
Cdkn2b |
AV083695, INK4b, MTS2, p15, p15(INK4b), p15INK4b |
-204.2kb | Anti-sense | Chr. 4 - 89306289 - 89311032 on NCBI Build GRCm38 |
| Downstream | doublesex and mab-3 related transcription factor like family A1 | Dmrta1 | Dmrt4 | 172.5kb | Sense | Chr. 4 - 89688198 - 89694766 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm12610
Genomic Map