Assay Details
| Gene Symbol: Zar1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159693.1 | NP_001153165.1 | 2 - 3 | 660 | 70 | |
| GenBank mRNA | FJ858201.1 | - | 2 - 3 | 660 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zar1l
Genomic Map