Assay Details
| Gene Symbol: Ccdc138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162956.1 | NP_001156428.1 | 9 - 10 | 1010 | 83 | |
| GenBank mRNA | BC119028.1 | - | 9 - 10 | 999 | 8733777 | 83 |
| BC119030.1 | - | 9 - 10 | 999 | 8733779 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc138
Genomic Map