Assay Details
| Gene Symbol: Stx11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163590.1 | NP_001157062.1 | 1 - 2 | 261 | 55 | |
| GenBank mRNA | AK171040.1 | - | 1 - 2 | 179 | 55 | |
| AK171663.1 | - | 187 | 55 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK171040.1, AK171663.1, NM_001163590.1 transcripts.
Set Membership: |
|

Stx11
Genomic Map