Assay Details
| Gene Symbol: Apol6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163621.1 | NP_001157093.1 | 2 - 3 | 187 | 64 | |
| NM_028010.1 | NP_082286.1 | 2 - 3 | 155 | 64 | ||
| GenBank mRNA | AK010208.1 | - | 2 - 3 | 156 | 64 | |
| AK144563.1 | - | 2 - 3 | 188 | 64 | ||
| BC113126.1 | - | 2 - 3 | 533 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK010208.1, NM_001163621.1, NM_028010.1 transcripts.
Set Membership: |
|

Apol6
Genomic Map