Assay Details
| Gene Symbol: Phf11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164289.1 | NP_001157761.1 | 3 - 4 | 722 | 81 | |
| GenBank mRNA | EU155106.1 | - | 3 - 4 | 2592 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phf11c
Genomic Map