Assay Details
| Gene Symbol: Rps27l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026467.3 | NP_080743.1 | 2 - 3 | 171 | 84 | |
| GenBank mRNA | AK007689.1 | - | 2 - 3 | 172 | 84 | |
| AK011262.1 | - | 2 - 3 | 190 | 84 | ||
| AK151083.1 | - | 2 - 3 | 190 | 84 | ||
| BC058115.1 | - | 2 - 3 | 172 | 6407738 | 84 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rps27l
Genomic Map