Assay Details
| Gene Symbol: Zswim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027198.1 | NP_081474.1 | 1 - 2 | 141 | 85 | |
| GenBank mRNA | AK010472.1 | - | 1 - 2 | 121 | 85 | |
| BC109343.2 | - | 1 - 2 | 99 | 40053371 | 85 | |
| BQ554850.1 | - | 1 - 2 | 144 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zswim7
Genomic Map