Assay Details
| Gene Symbol: Zswim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027198.1 | NP_081474.1 | 3 - 4 | 263 | 100 | |
| GenBank mRNA | AK010472.1 | - | 3 - 4 | 243 | 100 | |
| BC109343.2 | - | 3 - 4 | 221 | 40053371 | 100 | |
| BQ554850.1 | - | 3 - 4 | 266 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zswim7
Genomic Map