Assay Details
| Gene Symbol: Zufsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028287.2 | NP_082563.1 | 6 - 7 | 1173 | 104 | |
| GenBank mRNA | AK046191.1 | - | 6 - 7 | 1176 | 104 | |
| AK052071.1 | - | 6 - 7 | 1157 | 104 | ||
| AK077304.1 | - | 6 - 7 | 1221 | 104 | ||
| AK077730.1 | - | 6 - 7 | 1194 | 104 | ||
| AK160928.1 | - | 6 - 7 | 1172 | 104 | ||
| BC089514.1 | - | 6 - 7 | 1186 | 30252639 | 104 | |
More Information
Set Membership: |
|

Zufsp
Genomic Map