Assay Details
| Gene Symbol: Zufsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028287.2 | NP_082563.1 | 7 - 8 | 1360 | 109 | |
| GenBank mRNA | AK046191.1 | - | 7 - 8 | 1363 | 109 | |
| AK052071.1 | - | 7 - 8 | 1344 | 109 | ||
| AK077304.1 | - | 7 - 8 | 1408 | 109 | ||
| AK077730.1 | - | 7 - 8 | 1381 | 109 | ||
| AK160928.1 | - | 7 - 8 | 1359 | 109 | ||
| AK164346.1 | - | 4 - 5 | 2835 | 109 | ||
| AK164710.1 | - | 4 - 5 | 2706 | 109 | ||
| BC067065.1 | - | 5 - 6 | 1105 | 109 | ||
| BC089514.1 | - | 7 - 8 | 1373 | 30252639 | 109 | |
More Information
Set Membership: |
|

Zufsp
Genomic Map