Assay Details
| Gene Symbol: Zufsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028287.2 | NP_082563.1 | 9 - 10 | 1673 | 88 | |
| GenBank mRNA | AK046191.1 | - | 9 - 10 | 1676 | 88 | |
| AK052071.1 | - | 9 - 10 | 1657 | 88 | ||
| AK077730.1 | - | 9 - 10 | 1694 | 88 | ||
| AK160928.1 | - | 9 - 10 | 1672 | 88 | ||
| AK164346.1 | - | 6 - 7 | 3148 | 88 | ||
| AK164710.1 | - | 6 - 7 | 3019 | 88 | ||
| AK169467.1 | - | 2 - 3 | 335 | 88 | ||
| BC067065.1 | - | 7 - 8 | 1418 | 88 | ||
| BC089514.1 | - | 9 - 10 | 1686 | 30252639 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zufsp
Genomic Map