Assay Details
| Gene Symbol: 9430038I01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029886.2 | NP_084162.1 | 1 - 2 | 120 | 73 | |
| GenBank mRNA | AK020460.1 | - | 1 - 2 | 92 | 73 | |
| AK051763.1 | - | 1 - 2 | 96 | 73 | ||
| AK078159.1 | - | 1 - 2 | 100 | 73 | ||
| AK171836.1 | - | 1 - 2 | 121 | 73 | ||
| BY148312.1 | - | 1 - 2 | 120 | 73 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9430038I01Rik
Genomic Map