Assay Details
| Gene Symbol: Tmem161a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145597.4 | NP_663572.1 | 1 - 2 | 33 | 88 | |
| NR_122028.1 | 1 - 2 | 33 | 88 | |||
| GenBank mRNA | AK032711.1 | - | 1 - 2 | 37 | 88 | |
| AK049979.1 | - | 1 - 2 | 32 | 88 | ||
| AK082758.1 | - | 1 - 2 | 29 | 88 | ||
| AK147962.1 | - | 1 - 2 | 26 | 88 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | myocyte enhancer factor 2B | Mef2b | AI451606 | -4.9kb | Sense | Chr. 8 - 70152778 - 70167488 on NCBI Build GRCm38 |
| Downstream |
solute carrier family 25, member 42 |
Slc25a42 | 2900084M01Rik | 9.9kb | Anti-sense | Chr. 8 - 70184340 - 70212281 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK032711.1, AK049979.1, AK147962.1, NM_145597.4 transcripts.
Set Membership: |
|

Tmem161a
Genomic Map