Assay Details
| Gene Symbol: Hcar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175520.4 | NP_780729.2 | 1 | 1347 | 64 | |
| GenBank mRNA | AK029064.1 | - | 1 | 1332 | 64 | |
| AK046700.1 | - | 1 | 1349 | 64 | ||
| BC141305.1 | - | 1 | 1218 | 9055917 | 64 | |
| EU809459.1 | - | 1 | 953 | 64 | ||
| EU809460.1 | - | 1 | 929 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hcar1
Genomic Map