Assay Details
| Gene Symbol: AU022252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012400.2 | NP_001012400.1 | 5 | 1553 | 93 | |
| GenBank mRNA | AK158349.1 | - | 5 | 1512 | 93 | |
| BC035522.1 | - | 5 | 1536 | 93 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23244383 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23244379 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23244378 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23244381 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23244382 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2141311 | AK158349.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2141311 | AK167445.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2141311 | BC035522.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2141311 | BC089614.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2141311 | NM_001012400.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 498639 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 331448 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59012 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 869788 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 458782 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 162610 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130690 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2141311 | AK158349.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2141311 | AK167445.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2141311 | BC035522.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2141311 | BC089614.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2141311 | NM_001012400.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23244383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23244375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23244374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23244381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23244361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23244367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23244369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23244384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23244356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23244362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23244371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23244373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23244364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23244377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23244359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23244380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23244372 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130690 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00970816 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00970810 | - | - |
| Whole Mouse Genome G4122A | A_52_P347847 | BC035522.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P347847 | BC055790.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130690 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04008295 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03436506 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04008308 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04008287 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23244382 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23244381 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23244383 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23244379 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23244378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23244311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23244371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23244373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23244374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23244356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23244367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23244362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03830186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03758795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03709524 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23244346 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23244351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03735619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23244355 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23244348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23244372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03715901 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04008287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03573880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23244380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23244361 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23244377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23244354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23244375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03735626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23244365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03735620 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10515875 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515873 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515871 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515874 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515876 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515872 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438226_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10515870 | - | - |
| Mouse Genome 430 2.0 Array | 1438226_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438226_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 230696_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 230696_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400027693.mm.1 | TC0400004221.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400027693.mm.1 | NM_001012400.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027693.mm.1 | NM_001012400.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027695.mm.1 | TC0400004221.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400027695.mm.1 | NM_001012400.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027695.mm.1 | NM_001012400.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027700.mm.1 | TC0400004221.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400027700.mm.1 | NM_001012400.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027700.mm.1 | NM_001012400.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027703.mm.1 | TC0400004221.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400027703.mm.1 | NM_001012400.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027703.mm.1 | NM_001012400.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027694.mm.1 | TC0400004221.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400027694.mm.1 | NM_001012400.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027694.mm.1 | NM_001012400.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027692.mm.1 | TC0400004221.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400027692.mm.1 | NM_001012400.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027692.mm.1 | NM_001012400.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400035206.mm.1 | TC0400004221.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400035206.mm.1 | NM_001012400.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400035206.mm.1 | NM_001012400.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027702.mm.1 | TC0400004221.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400027702.mm.1 | NM_001012400.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027702.mm.1 | NM_001012400.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027696.mm.1 | TC0400004221.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400027696.mm.1 | NM_001012400.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027696.mm.1 | NM_001012400.2 | Refseq |
| Mouse Expression Set 430 | 1438226_PM_at | - | - |
| Mouse Expression Set 430 | 1438226_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438226_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438226_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1249278 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2893437 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2893437 | - | - |

AU022252
Genomic Map