Assay Details
| Gene Symbol: Trappc3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162937.1 | NP_001156409.1 | 6 | 747 | 92 | |
| GenBank mRNA | BC099502.1 | - | 6 | 747 | 30938327 | 92 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987554 | BC099502.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987554 | NM_001162937.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 920936 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 207824 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 570585 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724756 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573829 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 653435 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 667655 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 330455 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527607 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 557648 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 461055 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 620333 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 266996 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 327277 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 502371 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773482 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 958138 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 561737 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 896655 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 377157 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 796481 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 850488 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 160634 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656218 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 600790 | AK035949.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 600790 | AK049881.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 600790 | BC035535.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 600790 | NM_175513.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303282 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947670 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789732 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 707935 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654263 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 104233 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416955 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 511058 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 463370 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830599 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 755558 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 293446 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668114 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190053 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 46901 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409482 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 624952 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580789 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 20600 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 951943 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 466312 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477382 | AK031165.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477382 | AK031774.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477382 | AK044755.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477382 | BC060684.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932493 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 39878 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 38164 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 14075 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 82761 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 67128 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 173972 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 14102 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 36036 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 87078 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12984 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 4150 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10507 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 27953 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160380 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107349 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987554 | BC099502.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987554 | NM_001162937.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02099598 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02099520 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07023750 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02099561 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07023740 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160380 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07023812 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107349 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02099615 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02099615 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107349 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02099520 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160380 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07023740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27055187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08407219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08427579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27055190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27055192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08658961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27055181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27055193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08427577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27055188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27055184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08658960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27055186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08219962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27054931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08407216 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10362481 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362480 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362483 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362485 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362486 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362482 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362484 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10362479 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000001975.mm.1 | NM_001162937.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001975.mm.1 | NM_001162937.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001975.mm.1 | TC1000000295.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000001972.mm.1 | NM_001162937.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001972.mm.1 | NM_001162937.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001972.mm.1 | TC1000000295.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000001971.mm.1 | NM_001162937.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001971.mm.1 | NM_001162937.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001971.mm.1 | TC1000000295.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000001973.mm.1 | NM_001162937.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001973.mm.1 | NM_001162937.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001973.mm.1 | TC1000000295.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000001974.mm.1 | NM_001162937.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001974.mm.1 | NM_001162937.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001974.mm.1 | TC1000000295.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000001976.mm.1 | NM_001162937.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001976.mm.1 | NM_001162937.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001976.mm.1 | TC1000000295.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000001970.mm.1 | NM_001162937.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001970.mm.1 | NM_001162937.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001970.mm.1 | TC1000000295.mm.1 | MTA TC |

Trappc3l
Genomic Map