Assay Details
| Gene Symbol: Gprasp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163015.1 | NP_001156487.1 | 7 | 3485 | 78 | |
| NM_001163016.1 | NP_001156488.1 | 7 | 3447 | 78 | ||
| NM_001163017.1 | NP_001156489.1 | 7 | 3432 | 78 | ||
| GenBank mRNA | AK030638.1 | - | 7 | 3425 | 78 | |
| BC117923.2 | - | 7 | 3451 | 40097504 | 78 | |
| BC117924.2 | - | 7 | 3401 | 40097505 | 78 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32678434 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004213 | AK030638.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004213 | NM_001163017.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004213 | NM_001163016.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004213 | NM_001163015.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 233202 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 128776 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 751810 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 163421 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144359 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004213 | AK030638.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004213 | NM_001163017.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004213 | NM_001163016.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004213 | NM_001163015.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144359 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144359 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32678434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678408 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678420 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14558966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15149636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678414 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15175946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678426 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14599609 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10601838 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601835 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601840 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601842 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601843 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601841 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601837 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601836 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434073_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10601834 | - | - |
| Mouse Genome 430 2.0 Array | 1434073_at | - | - |
| Mouse Genome 430 2.0 Array | 1434073_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 245607_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 245607_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00008228.mm.1 | NM_001163015.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008228.mm.1 | NM_001163015.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008228.mm.1 | NM_001163016.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008228.mm.1 | NM_001163016.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008228.mm.1 | NM_001163017.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008228.mm.1 | NM_001163017.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008228.mm.1 | TC0X00003401.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008241.mm.1 | NM_001163015.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008241.mm.1 | NM_001163015.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008241.mm.1 | NM_001163016.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008241.mm.1 | NM_001163016.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008241.mm.1 | NM_001163017.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008241.mm.1 | NM_001163017.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008241.mm.1 | TC0X00003401.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00021154.mm.1 | NM_001163015.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00021154.mm.1 | NM_001163015.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00021154.mm.1 | NM_001163016.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00021154.mm.1 | NM_001163016.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00021154.mm.1 | NM_001163017.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00021154.mm.1 | NM_001163017.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00021154.mm.1 | TC0X00003401.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008243.mm.1 | NM_001163015.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008243.mm.1 | NM_001163015.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008243.mm.1 | NM_001163016.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008243.mm.1 | NM_001163016.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008243.mm.1 | NM_001163017.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008243.mm.1 | NM_001163017.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008243.mm.1 | TC0X00003401.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008219.mm.1 | NM_001163015.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008219.mm.1 | NM_001163015.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008219.mm.1 | TC0X00003401.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008222.mm.1 | NM_001163015.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008222.mm.1 | NM_001163015.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008222.mm.1 | NM_001163016.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008222.mm.1 | NM_001163016.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008222.mm.1 | NM_001163017.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008222.mm.1 | NM_001163017.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008222.mm.1 | TC0X00003401.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008237.mm.1 | NM_001163015.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008237.mm.1 | NM_001163015.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008237.mm.1 | NM_001163016.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008237.mm.1 | NM_001163016.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008237.mm.1 | NM_001163017.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008237.mm.1 | NM_001163017.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008237.mm.1 | TC0X00003401.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008242.mm.1 | NM_001163015.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008242.mm.1 | NM_001163015.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008242.mm.1 | NM_001163016.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008242.mm.1 | NM_001163016.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008242.mm.1 | NM_001163017.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008242.mm.1 | NM_001163017.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008242.mm.1 | TC0X00003401.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008236.mm.1 | NM_001163015.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008236.mm.1 | NM_001163015.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008236.mm.1 | NM_001163016.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008236.mm.1 | NM_001163016.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008236.mm.1 | NM_001163017.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008236.mm.1 | NM_001163017.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008236.mm.1 | TC0X00003401.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008235.mm.1 | NM_001163015.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008235.mm.1 | NM_001163015.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008235.mm.1 | NM_001163016.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008235.mm.1 | NM_001163016.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008235.mm.1 | NM_001163017.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008235.mm.1 | NM_001163017.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008235.mm.1 | TC0X00003401.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008217.mm.1 | NM_001163015.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008217.mm.1 | NM_001163015.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008217.mm.1 | NM_001163016.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008217.mm.1 | NM_001163016.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008217.mm.1 | NM_001163017.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008217.mm.1 | NM_001163017.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008217.mm.1 | TC0X00003401.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008232.mm.1 | NM_001163015.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008232.mm.1 | NM_001163015.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008232.mm.1 | NM_001163016.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008232.mm.1 | NM_001163016.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008232.mm.1 | NM_001163017.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008232.mm.1 | NM_001163017.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008232.mm.1 | TC0X00003401.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434073_at | - | - |
| Mouse Expression Set 430 | 1434073_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434073_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434073_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2730909 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2730909 | - | - |

Gprasp2
Genomic Map