Assay Details
Gene Symbol: Gm17689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167584.1 | NP_001161056.1 | 1 - 2 | 93 | 92 | |
GenBank mRNA | EU703627.1 | - | 1 - 2 | 93 | 92 |

More Information
Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0800000438.rn
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
